4-Hydroxythiophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR4561-25G OR4561-5G OR4561-100G
76611-888EA 51.96 USD
76611-888 76611-890 76611-886
4-Hydroxythiophenol 98%
4-Hydroxythiophenol
Formula: C₆H₆OS
Boiling Pt: 149…150 °C/25mmHg
Melting Pt: 33…35 °C
Density: 1.255 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004850
CAS Number: 637-89-8
EINECS: 211-307-6
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR