(R)-(+)-2-Chloropropionic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR52411-100G OR52411-25G OR52411-5G
76612-016EA 132.4 USD
76612-016 76612-018 76612-020
(R)-(+)-2-Chloropropionic acid 98%
(R)-(+)-2-Chloropropionic acid
Formula: H₃CCHClCOOH
Melting Pt: –12…0 °C
Density: 1.258 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00064204
CAS Number: 7474-05-7
UN: 2511
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR