Copper(I) chloride 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Cuprous chloride, Copper chloride

IN1542-100G IN1542-1KG IN1542-500G
76610-776EA 44.46 USD
76610-776 76610-778 76610-780
Copper(I) chloride 99%
Copper (I) chloride
Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
Density: 4.14 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010971
CAS Number: 7758-89-6
EINECS: 231-842-9
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR