Indole

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1H-Indole

OR10189-100G OR10189-25G OR10189-500G
76615-308EA 41.76 USD
76615-308 76615-310 76615-312
Indole
Indole
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253…254 °C
Melting Pt: 52…54 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,04985

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR