Dibenzofuran

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Dibenzo[b,d]furan

OR5267-100G OR5267-25G OR5267-50G
76616-946EA 41.76 USD
76616-946 76616-948 76616-950
Dibenzofuran
Dibenzofuran
Formula: C₁₂H₈O
MW: 168.19 g/mol
Boiling Pt: 285
Melting Pt: 80…82 °C
Density: 1.089 g/cm³ (25 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004968
CAS Number: 132-64-9
EINECS: 205-071-3
UN: 3077
ADR: 9+EHS III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR