1,2,3,4,5-Pentamethylcyclopentadiene, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR318024-1G OR318024-25G OR318024-5G
76625-122EA 20.32 USD
76625-122 76625-124 76625-126
1,2,3,4,5-Pentamethylcyclopentadiene, Technical Grade
1,2,3,4,5-Pentamethylcyclopentadiene
Formula: C₁₀H₁₆
Boiling Pt: 58 °C/15mmHg
Density: 0.87 g/cm³ (25 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001354
CAS Number: 4045-44-7
EINECS: 223-743-4
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR