Bromoacetonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR52127-100G OR52127-25G OR52127-500G
76625-262EA 69.26 USD
76625-262 76625-264 76625-266
Bromoacetonitrile 97%
Bromoacetonitrile
Formula: BrCNCH₂
MW: 119.95 g/mol
Boiling Pt: 150
Melting Pt: -36
Density: 1.722 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001884
CAS Number: 590-17-0
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR