Sodium iodide, anhydrous 99+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

IN3263-250G IN3263-100G IN3263-1KG
76615-108EA 69.26 USD
76615-108 76615-104 76615-106
Sodium iodide, anhydrous 99+%
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300
Melting Pt: 662
Density: 3.67 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9+EHS III
Merck Index: 13,08705

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR