4-Iodobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: p-Iodobenzonitrile

OR4305-100G OR4305-25G
76625-190EA 87.6 USD
76625-190 76625-192
4-Iodobenzonitrile
4-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Boiling Pt: 285
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00051310
CAS Number: 3058-39-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR