5-Chlorosalicylaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR1167-100G OR1167-25G
76615-484EA 45.84 USD
76615-484 76615-486 76615-488
5-Chlorosalicylaldehyde 97%
5-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR