2,4-Dibromophenol 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR13037-100G OR13037-25G
76615-594EA 55.02 USD
76615-594 76615-596
2,4-Dibromophenol 95%
2,4-Dibromophenol
Formula: C₆H₄Br₂O
MW: 251.91 g/mol
Boiling Pt: 239 °C
Melting Pt: 38
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002149
CAS Number: 615-58-7
EINECS: 210-436-5
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR