5-Bromoanthranilic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Amino-5-bromobenzoic acid

OR3626-100G OR3626-25G
76616-508EA 33.62 USD
76616-508 76616-510 76616-512
5-Bromoanthranilic acid 98%
5-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR