Zinc 98%, dust max. 10 μm

Supplier: APOLLO SCIENTIFIC
Warning

IN1013-100G IN1013-500G
76615-068EA 22.02 USD
76615-068 76615-070
Zinc 98%, dust max. 10 μm
Zinc

Delivery of this item to mainland UK is £40, additional charge will be added at order confirmation stage by our sales team.

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.133 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 3077
ADR: 9+EHS III
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR