4-Bromothiophene-3-boronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR8555-1G OR8555-250MG
76588-848EA 52.8 USD
76588-848 76588-846
4-Bromothiophene-3-boronic acid 97%
4-Bromothiophene-3-boronic acid
Formula: C₄H₄BBrO₂S
Melting Pt: 196…198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06801663
CAS Number: 101084-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR