Tetrakis(dimethylamino)diboron

Supplier: APOLLO SCIENTIFIC
Warning

OR7996-100G OR7996-25G OR7996-5G
76588-492EA 179.28 USD
76588-492 76588-490 76588-488
Tetrakis(dimethylamino)diboron
Tetrakis(dimethylamino)diboron
Formula: C₈H₂₄B₂N₄
Boiling Pt: 55…57 °C/2.5mmHg
Melting Pt: –33 °C
Density: 0.926 g/cm³ (25 °C)
Flash Pt: 98.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048012
CAS Number: 1630-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR