4-Bromo-9,9'-spirobi[9H-fluorene] 98%

Supplier: APOLLO SCIENTIFIC

OR51009-1G OR51009-25G OR51009-5G
76586-546EA 20.87 USD
76586-546 76586-550 76586-548
4-Bromo-9,9'-spirobi[9H-fluorene] 98%
4-Bromo-9,9'-spirobi[9H-fluorene]
Formula: C₂₅H₁₅Br
Storage Temperature: Ambient
MDL Number: MFCD16660920
CAS Number: 1161009-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR