Nortropinone hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 8-Azabicyclo[3.2.1]octan-3-one hydrochloride

OR4717-25G OR4717-5G
76585-424EA 48.9 USD
76585-424 76585-422
Nortropinone hydrochloride 97%
Nortropinone hydrochloride
Formula: C₇H₁₂ClNO
MW: 161.63 g/mol
Melting Pt: 204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03613582
CAS Number: 25602-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR