(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: XANTPHOS, 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

OR42230-100G OR42230-25G OR42230-5G
76583-494EA 184.38 USD
76583-494 76583-492 76583-490
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)
Formula: C₃₉H₃₂OP₂
MW: 578.62 g/mol
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00233866
CAS Number: 161265-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR