1-(Boc-amino)cyclopropanecarboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(Boc-Amino)cyclopropanecarboxylic acid, 1-Aminocyclopropane-1-carboxylic acid, N-BOC protected, 1-[(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic acid

OR8028-5G OR8028-1G OR8028-25G
76588-514EA 24.46 USD
76588-514 76588-512 76588-516
1-(Boc-amino)cyclopropanecarboxylic acid 97%
1-(Boc-amino)cyclopropanecarboxylic acid
Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Melting Pt: 177…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00083257
CAS Number: 88950-64-5
EINECS: 230-181-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR