tert-Butyl-5-(benzyloxy)-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-(Benzyloxy)-3-(hydroxymethyl)-1H-indole, N-BOC protected

OR1705-100MG OR1705-250MG OR1705-500MG
76568-518EA 20.77 USD
76568-518 76568-520 76568-522
tert-Butyl-5-(benzyloxy)-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%
tert-Butyl-5-(benzyloxy)-3-(hydroxymethyl)-1H-indole-1-carboxylate
Formula: C₂₁H₂₃NO₄
Storage Temperature: Ambient
CAS Number: 914349-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR