α-Methylcinnamic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methyl-3-phenyl-2-propenoic acid

OR911424-100G OR911424-25G
76589-566EA 72.32 USD
76589-566 76589-564
α-Methylcinnamic acid 97%
α-Methylcinnamic acid
Formula: C₁₀H₁₀O₂
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002652
CAS Number: 1199-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR