3-Hydroxytetrahydropyran

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Hydroxy-2H-pyran, Tetrahydro-2H-pyran-3-ol

OR46583-1G OR46583-250MG
76585-126EA 24.46 USD
76585-126 76585-124 76585-128
3-Hydroxytetrahydropyran
3-Hydroxytetrahydropyran
Formula: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00219387
CAS Number: 19752-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR