Pyromellitic dianhydride 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR4903-100G OR4903-2.5KG OR4903-500G
76585-874EA 20.97 USD
76585-874 76585-878 76585-876
Pyromellitic dianhydride 98%
Pyromellitic dianhydride
Formula: C₁₀H₂O₆
Boiling Pt: 397…400 °C
Melting Pt: 283…288 °C
Density: 1.68 g/cm³ (25 °C)
Flash Pt: 380 °C
Storage Temperature: Ambient
MDL Number: MFCD00005005
CAS Number: 89-32-7
EINECS: 201-898-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR