N,N-Dimethylglycine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

OR45130-5G OR45130-100G OR45130-25G
76584-258EA 20.87 USD
76584-258 76584-262 76584-260
N,N-Dimethylglycine 97%
N,N-Dimethylglycine
Formula: C₄H₉NO₂
Melting Pt: 178…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR