Indole-3-carbaldehyd 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde

OR5052-100G OR5052-250G OR5052-500G
76586-220EA 33.62 USD
76586-220 76586-222 76586-224
Indole-3-carbaldehyd 99%
Indole-3-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 190…199 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00005622
CAS Number: 487-89-8
EINECS: 207-665-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR