3-Bromo-7-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC
Danger

OR6862T-1G OR6862T-5G
76587-182EA 20.26 USD
76587-182 76587-180 76587-184
3-Bromo-7-nitro-1H-indazole
3-Bromo-7-nitro-1H-indazole

A more potent inhibitor of rat cerebellar nitric oxide synthase than 7-nitroindazole.Harmful to reproduction.

Formula: C₇H₄BrN₃O₂
Melting Pt: 177…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00159910
CAS Number: 74209-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR