Uracil

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Pyrimidine-2,4(1H,3H)-dione

OR4885-100G OR4885-250G OR4885-25G
76585-860EA 34.64 USD
76585-860 76585-862 76585-858
Uracil
Uracil
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR