Tetraoctylammonium bromide

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tetra-n-octylammonium bromide

OR4675-100G OR4675-25G OR4675-500G
76585-212EA 24.46 USD
76585-212 76585-210 76585-214
Tetraoctylammonium bromide
Tetraoctylammonium bromide
Formula: C₃₂H₆₈BrN
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00011863
CAS Number: 14866-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR