4-Bromo-3-methylanisole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene

OR4933-10G OR4933-25G
76585-918EA 20.89 USD
76585-918 76585-920 76585-916
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole
Formula: C₈H₉BrO
Boiling Pt: 111…112 °C/15mmHg
Density: 1.424 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00060687
CAS Number: 27060-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR