9-Fluorenylmethanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol

OR7130-100G OR7130-1KG OR7130-25G
76587-690EA 23.42 USD
76587-690 76587-692 76587-688
9-Fluorenylmethanol
9-Fluorenylmethanol
Formula: C₁₄H₁₂O
Melting Pt: 102…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001139
CAS Number: 24324-17-2
EINECS: 246-167-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR