Naphthacene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3-Benzanthracene, Benz[b]anthracene, Tetracene

OR50973-1G OR50973-250MG
76586-340EA 151.78 USD
76586-340 76586-338
Naphthacene
Naphthacene
Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 437
Melting Pt: 356
Storage Temperature: Ambient
MDL Number: MFCD00003702
CAS Number: 92-24-0
Merck Index: 12,06456

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR