Tetrabutylammonium hydrogen sulfate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulfate, Tetrabutylammonium bisulfate, Tetrabutylammonium hydrogensulphate

OR4670-1KG OR4670-500G OR4670-5KG
76585-180EA 143.52 USD
76585-180 76585-178 76585-182
Tetrabutylammonium hydrogen sulfate 97%
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
Melting Pt: 168…172 °C
Density: 1.01 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR