4-Methoxypyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR8764-25G OR8764-5G
76589-056EA 24.46 USD
76589-056 76589-054 76589-058
4-Methoxypyridine
4-Methoxypyridine
Formula: C₆H₇NO
Boiling Pt: 191 °C
Melting Pt: 4 °C
Density: 1.075 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674049
CAS Number: 620-08-6
EINECS: 210-624-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR