3,4-Dimethoxyaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR4959-25G OR4959-500G OR4959-100G
76585-946EA 20.87 USD
76585-946 76585-950 76585-948
3,4-Dimethoxyaniline 98%
3,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
Boiling Pt: 174…176 °C/22mmHg
Melting Pt: 85…89 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR