2,5-Dibromobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR4523-25G OR4523-5G
76584-704EA 44 USD
76584-704 76584-702
2,5-Dibromobenzoic acid
2,5-Dibromobenzoic acid
Formula: C₇H₄Br₂O₂
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00016494
CAS Number: 610-71-9
EINECS: 210-234-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR