9-Bromoanthracene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR8575-25G OR8575-5G
76588-882EA 121.54 USD
76588-882 76588-880 76588-878
9-Bromoanthracene 98%
9-Bromoanthracene
Formula: C₁₄H₉Br
Melting Pt: 97…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR