Methyl 2-bromo-3-chloro-6-fluorobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

PC501762-1G PC501762-250MG
76560-488EA 146.52 USD
76560-488 76561-810
Methyl 2-bromo-3-chloro-6-fluorobenzoate
Methyl 2-bromo-3-chloro-6-fluorobenzoate
Formula: C₈H₅BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28141930
CAS Number: 1805210-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR