1-(2-Fluorophenyl)biguanide hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4149G-1G PC4149G-25G PC4149G-5G
76562-998EA 48.26 USD
76562-998 76560-412 76561-476
1-(2-Fluorophenyl)biguanide hydrochloride 97%
1-(2-Fluorophenyl)biguanide hydrochloride
Formula: C₈H₁₁ClFN₅
Melting Pt: 237…241 °C
Storage Temperature: Ambient
MDL Number: MFCD00236682
CAS Number: 66088-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR