4-Chloro-2-(trifluoromethyl)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane

PC1777-100G PC1777-25G PC1777-5G
76559-800EA 399.32 USD
76559-800 76559-980 76560-160
4-Chloro-2-(trifluoromethyl)phenylboronic acid 98%
4-Chloro-2-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD04973086
CAS Number: 313545-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR