Boc-trans-4-hydroxy-L-proline 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline
OR307069-25G
OR307069-100G
76574-742EA
23.34
USD
76574-742
76574-744
76574-746
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline
Formula:
C₁₀H₁₇NO₅ MW: 231.3 g/mol Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053370 CAS Number: 13726-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...