Bis(triphenylphosphine)palladium(II) chloride 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Bis(triphenylphosphine)palladium(II) dichloride
OR30692-25G
OR30692-1G
OR30692-5G
76574-702EA
552.12
USD
76574-702
76574-698
76574-700
Bis(triphenylphosphine)palladium(II) chloride 98%
Bis(triphenylphosphine)palladium (II) chloride
Formula:
C₃₆H₃₀Cl₂P₂Pd MW: 701.91 g/mol Melting Pt: 310 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009593 CAS Number: 13965-03-2 EINECS: 237-744-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...