2,2'-Bipyridine-4,4'-dicarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid

OR301064-5G OR301064-10G OR301064-1G
76573-662EA 34.64 USD
76573-662 76573-664 76573-660
2,2'-Bipyridine-4,4'-dicarboxylic acid
2,2'-Bipyridine-4,4'-dicarboxylic acid
Formula: C₁₂H₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00015430
CAS Number: 6813-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR