2-Chloro-1H-indole-3-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR29092-5G OR29092-1G
76572-826EA 135.48 USD
76572-826 76572-824
2-Chloro-1H-indole-3-carbaldehyde
2-Chloro-1H-indole-3-carbaldehyde
Formula: C₉H₆ClNO
MW: 179.61 g/mol
Melting Pt: 211…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00111620
CAS Number: 5059-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR