(-)-Quinine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

OR28880-25G OR28880-1G OR28880-5G
76572-744EA 44.82 USD
76572-744 76572-740 76572-742
(-)-Quinine
(-)-Quinine
Formula: C₂₀H₂₄N₂O₂
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00198096
CAS Number: 130-95-0
EINECS: 205-003-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR