2-Adamantanol

Supplier: APOLLO SCIENTIFIC

OR28840-25G OR28840-5G
76572-712EA 74.38 USD
76572-712 76572-710
2-Adamantanol
2-Adamantanol
Formula: C₁₀H₁₆O
Melting Pt: 258…262 °C
Storage Temperature: Ambient
MDL Number: MFCD00074744
CAS Number: 700-57-2
EINECS: 211-846-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR