Disodium-1,6-naphthalenedisulfonate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,6-Naphthalenedisulphonic acid Disodium salt, 1,6-Naphthalenedisulfonic acid Disodium salt, Sodium naphthalene-1,6-disulphonate, Sodium naphthalene-1,6-disulfonate

OR2838-1KG OR2838-500G OR2838-100G
76572-396EA 107.98 USD
76572-396 76572-394 76572-392
Disodium-1,6-naphthalenedisulfonate 97%
Disodium-1,6-naphthalenedisulfonate
Formula: C₁₀H₈O₆S₂
MW: 332.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021522
CAS Number: 1655-43-2
EINECS: 216-734-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR