2-(4-Chlorophenyl)ethanol 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chlorophenethyl alcohol

OR24970-100G OR24970-25G
76571-518EA 31.38 USD
76571-518 76571-516 76571-514
2-(4-Chlorophenyl)ethanol 99%
2-(4-Chlorophenyl)ethanol
Formula: C₈H₉ClO
Boiling Pt: 110 °C/0.5mmHg
Density: 1.157 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002899
CAS Number: 1875-88-3
EINECS: 217-506-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR