2-Nitro-1-naphthol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Hydroxy-2-nitronaphthalene

OR24530-1G OR24530-5G
76571-404EA 20.77 USD
76571-404 76571-406
2-Nitro-1-naphthol 98%
2-Nitro-1-naphthol
Formula: C₁₀H₇NO₃
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003956
CAS Number: 607-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR