Chitin

Supplier: APOLLO SCIENTIFIC
Warning

OR2403-250G OR2403-5G OR2403-25G
76571-232EA 126.32 USD
76571-232 76571-228 76571-230
Chitin
Chitin
Formula: C₃₀H₅₀N₄O₁₉
MW: 233.26 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00466914
CAS Number: 1398-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR