4-Phenylbutyric acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Phenylbutanoic acid

OR2390-500G OR2390-100G OR2390-25G
76571-194EA 309.68 USD
76571-194 76571-192 76571-190
4-Phenylbutyric acid 98%
4-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
Boiling Pt: 165 °C/10mmHg
Melting Pt: 49…53 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR